Abnova
Manufacturer Code:PAB14167
Catalog # 89-116-967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human MARCKS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MARCKS.
For Research Use Only
Protein Aliases: 80K-L; 80K-L protein; FLJ14368; FLJ90045; MACS; MARCKS; Myristoylated alanine-rich C-kinase substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; Protein kinase C substrate, 80 kDa protein, light chain
Gene Aliases: 80K-L; MACS; MARCKS; PKCSL; PRKCSL
UniProt ID: (Human) P29966
Entrez Gene ID: (Human) 4082
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.