OriGene
Manufacturer Code:CF809144
Catalog # 50-166-9301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6G3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 623-758 of human MAPT produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DDPAC FTDP-17 MAPTL MSTD MTBT1 MTBT2 PPND PPP1R103 TAU; G protein beta1/gamma2 subunit-interacting factor 1; Microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; Neurofibrillary tangle protein; Paired helical filament-tau; PHF-tau; protein phosphatase 1, regulatory subunit 103
Gene Aliases: DDPAC; FTDP-17; MAPT; MAPTL; MSTD; MTBT1; MTBT2; PPND; PPP1R103; TAU
UniProt ID: (Human) P10636
Entrez Gene ID: (Human) 4137
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.