Novus Biologicals
Manufacturer Code:NBP19836620UL
Catalog # NBP19836620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Mapre1 - C-terminal region. Peptide sequence GKLRNIELICQENEGENDPVLQRIVDILYATDEGFVIPDEGGPQEEQEEY. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenomatous polyposis coli-binding protein EB1; adenomatous polyposis coli-binding protein EB1 APC-binding protein EB1 EB1MGC129946 End-binding protein 1 MGC117374 microtubule-associated protein RP/EB family member 1 microtubule-associated protein RP/EB family member 1; APC-binding protein EB1; EB1; End-binding protein 1; Microtubule-associated protein RP/EB family member 1; microtubule-associated protein, RP/EB family, member 1
Gene Aliases: EB1; MAPRE1
UniProt ID: (Human) Q15691
Entrez Gene ID: (Human) 22919
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.