Invitrogen
Manufacturer Code:PA576477
Catalog # PIPA576477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide of human MAPKBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JNK-binding protein 1; JNKBP-1; Jun N-terminal kinase binding protein 1; Jun N-terminal kinase-binding protein 1; mitogen activated protein kinase binding protein 1; Mitogen-activated protein kinase-binding protein 1; Mitogen-activated protein kinase-binding protein 1 JNK-binding protein 1 JNKBP-1 JNKBP1 KIAA0596
Gene Aliases: 2810483F24Rik; AW123212; JNKBP-1; JNKBP1; KIAA0596; MAPKBP1; mKIAA0596
UniProt ID: (Human) O60336, (Mouse) Q6NS57
Entrez Gene ID: (Human) 23005, (Mouse) 26390, (Rat) 362197
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.