Novus Biologicals
Manufacturer Code:NBP229677
Catalog # NBP229677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9F10.2 |
Immunogen | KLH-conjugated synthetic peptide containing the sequence and surrounding amino acids phosphorylated Thr334 of human mouse rat bovine and chicken MAPKAP Kinase 2. Immunizing peptide shared 66% homology with phospho-MAPKAP-K3 at Thr313. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 MAP kinase-activated protein kinase 2 MAPK-activated protein kinase 2 MAPKAP kinase 2 MAPKAPK-2 mitogen-activated protein kinase-activated protein kinase 2 MK2; MAP kinase-activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2
Gene Aliases: MAPKAP-K2; MAPKAPK2; MK-2; MK2
UniProt ID: (Human) P49137
Entrez Gene ID: (Human) 9261
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.