Novus Biologicals
Manufacturer Code:NBP258247
Catalog # NBP258247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TMRVDYEQIKIKKIEDASNPLLLKRRKKARALEAAAL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 MAP kinase-activated protein kinase 2 MAPK-activated protein kinase 2 MAPKAP kinase 2 MAPKAPK-2 mitogen-activated protein kinase-activated protein kinase 2 MK2; MAP kinase-activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2
Gene Aliases: MAPKAP-K2; MAPKAPK2; MK-2; MK2
UniProt ID: (Human) P49137
Entrez Gene ID: (Human) 9261
Molecular Function: cytoskeletal protein kinase microtubule family cytoskeletal protein non-motor microtubule binding protein non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.