Abnova
Manufacturer Code:PAB17382
Catalog # 89-119-038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human MAPK8IP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MAPK8IP2.
For Research Use Only
Protein Aliases: C-Jun-amino-terminal kinase-interacting protein 2; homologous to mouse JIP-1; IB-2; IB2; islet-brain 2; Islet-brain-2; JIP-2; JIP2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; Mitogen-activated protein kinase 8-interacting protein 2; PRKM8IPL
Gene Aliases: IB-2; IB2; JIP2; MAPK8IP2; PRKM8IPL
UniProt ID: (Human) Q13387
Entrez Gene ID: (Human) 23542
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.