OriGene
Manufacturer Code:CF502150
Catalog # 50-166-1082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D4 |
Immunogen | Full length human recombit protein of human MAPK7 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Big MAP kinase 1; BMK-1; BMK1 ERK4 ERK5 PRKM7; BMK1 kinase; ERK-5; Extracellular signal-regulated kinase 5; extracellular-signal-regulated kinase 5; MAP kinase 7; MAPK 7; Mitogen-activated protein kinase 7
Gene Aliases: BMK1; ERK4; ERK5; MAPK7; PRKM7
UniProt ID: (Human) Q13164
Entrez Gene ID: (Human) 5598
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.