Novus Biologicals
Manufacturer Code:NBP158850
Catalog # NBP158850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MAP4K5(mitogen-activated protein kinase kinase kinase kinase 5) The peptide sequence was selected from the middle region of MAP4K5. Peptide sequence QGKSFKSDEVTQEISDETRVFRLLGSDRVVVLESRPTENPTAHSNLYILA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 GCKR germinal center kinase-related KHS1 KHSMEKKK 5 Kinase homologous to SPS1/STE20 MAPK/ERK kinase kinase kinase 5 MAPKKKK5 MEK kinase kinase 5 mitogen-activated protein kinase kinase kinase kinase 5; germinal center kinase-related; KHS; Kinase homologous to SPS1/STE20; MAPK/ERK kinase kinase kinase 5; MEK kinase kinase 5; MEKKK 5; Mitogen-activated protein kinase kinase kinase kinase 5
Gene Aliases: GCKR; KHS; KHS1; MAP4K5; MAPKKKK5
UniProt ID: (Human) Q9Y4K4
Entrez Gene ID: (Human) 11183
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.