Proteintech
Manufacturer Code:128271AP150UL
Catalog # 50-172-8280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAP4K5 Fusion Protein Ag3547 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCKR KHS KHS1 MAP4K5 MAPKKKK5 MEK kinase kinase 5 MEKKK 5; germinal center kinase-related; KHS; Kinase homologous to SPS1/STE20; map4k5; MAPK/ERK kinase kinase kinase 5; MEK kinase kinase 5; MEKKK 5; Mitogen-activated protein kinase kinase kinase kinase 5
Gene Aliases: 4432415E19Rik; GCKR; KHS; KHS1; MAP4K5; MAPKKKK5; RGD1562028
UniProt ID: (Human) Q9Y4K4, (Mouse) Q8BPM2
Entrez Gene ID: (Human) 11183, (Rat) 503027, (Mouse) 399510
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.