Abnova
Manufacturer Code:PAB8104
Catalog # 89-133-588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S629 of human MAP4K4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP4K4.
For Research Use Only
Protein Aliases: epididymis secretory protein Li 31; FLH21957; FLJ10410; FLJ20373; FLJ90111; hepatocyte progenitor kinase-like/germinal center kinase-like kinase; HGK; HPK/GCK-like kinase HGK; KIAA0687; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; Mitogen-activated protein kinase kinase kinase kinase 4; Nck-interacting kinase; NIK; Ste20 group protein kinase HGK
Gene Aliases: FLH21957; HEL-S-31; HGK; KIAA0687; MAP4K4; MEKKK4; NIK
UniProt ID: (Human) O95819
Entrez Gene ID: (Human) 9448
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.