Abnova
Manufacturer Code:H00009448A01
Catalog # 89-106-083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 611 a.a. &bsim 710 a.a) partial recombinant protein with GST tag.,MAP4K4 (NP_663719 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant MAP4K4.
For Research Use Only
Protein Aliases: epididymis secretory protein Li 31; FLH21957; FLJ10410; FLJ20373; FLJ90111; hepatocyte progenitor kinase-like/germinal center kinase-like kinase; HGK; HPK/GCK-like kinase HGK; KIAA0687; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; Mitogen-activated protein kinase kinase kinase kinase 4; Nck-interacting kinase; NIK; Ste20 group protein kinase HGK
Gene Aliases: FLH21957; HEL-S-31; HGK; KIAA0687; MAP4K4; MEKKK4; NIK
UniProt ID: (Human) O95819
Entrez Gene ID: (Human) 9448
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.