Abnova
Manufacturer Code:MAB12312
Catalog # 89-934-515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 219CT8.3.1 |
Immunogen | Recombinant protein corresponding to human MAP4K3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against human MAP4K3.
For Research Use Only
Protein Aliases: germinal center kinase-like kinase; Germinal center kinase-related protein kinase; GLK; MAPK/ERK kinase kinase kinase 3; MAPKKKK3; MEK kinase kinase 3; Mitogen-activated protein kinase kinase kinase kinase 3; RAB8IPL1
Gene Aliases: GLK; MAP4K3; MAPKKKK3; MEKKK 3; MEKKK3; RAB8IPL1
UniProt ID: (Human) Q8IVH8
Entrez Gene ID: (Human) 8491
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.