Novus Biologicals
Manufacturer Code:NBP30515420UL
Catalog # NB093523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 755-894 of human MAP4K3 (NP_003609.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 germinal center kinase-like kinase Germinal center kinase-related protein kinase GLKMEKKK 3 MAPK/ERK kinase kinase kinase 3 MAPKKKK3 MEK kinase kinase 3 mitogen-activated protein kinase kinase kinase kinase 3 RAB8IPL1; germinal center kinase-like kinase; Germinal center kinase-related protein kinase; GLK; MAPK/ERK kinase kinase kinase 3; MEK kinase kinase 3; Mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases: GLK; MAP4K3; MAPKKKK3; MEKKK 3; MEKKK3; RAB8IPL1
UniProt ID: (Human) Q8IVH8
Entrez Gene ID: (Human) 8491
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.