Invitrogen
Manufacturer Code:PA571682
Catalog # PIPA571682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 321-349 amino acids from the Central region of human MAP4K2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine-protein kinase; GC kinase; Germinal center kinase; germinal centre kinase (GC kinase); MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; Mitogen-activated protein kinase kinase kinase kinase 2; Mitogen-activated protein kinase kinase kinase kinase 2 B lymphocyte serine/threonine-protein kinase Germinal center kinase GC kinase MAPK/ERK kinase kinase kinase 2 MEK kinase kinase 2 MEKKK 2 Rab8-interacting protein MAP4K2 GCK RAB8IP; Rab8 interacting protein; Rab8-interacting protein
Gene Aliases: BL44; GCK; MAP4K2; RAB8IP
UniProt ID: (Human) Q12851
Entrez Gene ID: (Human) 5871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.