Novus Biologicals
Manufacturer Code:NBP188584
Catalog # NBP188584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ETDPLNEPWEEEWTLLGKEELSGSLLQSVQEALEERSLTIRSASEFQELDSPDDTMGTIKRAPFLGPLPTDPPAEEPLSSPPGTLPPPPSGPNSSPLLPTAWATMKQRE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine protein kinase B lymphocyte serine/threonine-protein kinase BL44 EC 2.7.11 EC 2.7.11.1 GC kinase GCKMEK kinase kinase 2 Germinal center kinase germinal centre kinase (GC kinase) MAPK/ERK kinase kinase kinase 2 mitogen-activated protein kinase kinase kinase kinase 2 Rab8 interacting protein Rab8-interacting protein RAB8IPMEKKK 2; B lymphocyte serine/threonine-protein kinase; GC kinase; Germinal center kinase; germinal centre kinase (GC kinase); MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; Mitogen-activated protein kinase kinase kinase kinase 2; Rab8 interacting protein; Rab8-interacting protein
Gene Aliases: BL44; GCK; MAP4K2; RAB8IP
UniProt ID: (Human) Q12851
Entrez Gene ID: (Human) 5871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.