Novus Biologicals
Manufacturer Code:NBP287775
Catalog # NB035397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of mouse MAP3K9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.25 MEKK9 mitogen-activated protein kinase kinase kinase 9 MLK1Mixed lineage kinase 1 PRKE1mixed lineage kinase 1 (tyr and ser/thr specificity); Mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity)
Gene Aliases: MAP3K9; MEKK9; MLK1; PRKE1
UniProt ID: (Human) P80192
Entrez Gene ID: (Human) 4293
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.