Abnova
Manufacturer Code:PAB14284
Catalog # 89-116-983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human MAP3K11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MAP3K11.
For Research Use Only
Protein Aliases: MAP3K11; MGC17114; Mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; MLK-3; MLK3; protein-tyrosine kinase PTK1; PTK1; SH3 domain-containing proline-rich kinase; SPRK; Src-homology 3 domain-containing proline-rich kinase
Gene Aliases: MAP3K11; MEKK11; MLK-3; MLK3; PTK1; SPRK
UniProt ID: (Human) Q16584
Entrez Gene ID: (Human) 4296
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.