Abnova
Manufacturer Code:PAB29207
Catalog # 89-130-641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T1400 of human MAP3K1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAP3K1.
For Research Use Only
Protein Aliases: MAP/ERK kinase kinase 1; MAPK/ERK kinase kinase 1; MAPKKK1; MEK kinase 1; MEKK; MEKK1; Mitogen-activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase
Gene Aliases: MAP3K1; MAPKKK1; MEKK; MEKK 1; MEKK1; SRXY6
UniProt ID: (Human) Q13233
Entrez Gene ID: (Human) 4214
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.