OriGene
Manufacturer Code:TA500411
Catalog # 50-167-1546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B4 |
Immunogen | Full-length protein expressed in 293T cell transfected with human MAP2K4 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-Jun N-terminal kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 4; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK JNKK1 MAPKK4 MEK4 MKK4 PRKMK4 SAPKK-1 SAPKK1 SEK1 SERK1 SKK1; MAP kinase kinase 4; MAPK/ERK kinase 4; MAPKK 4; MEK 4; SAPK kinase 1; SAPK/ERK kinase 1; SEK1; Stress-activated protein kinase kinase 1
Gene Aliases: JNKK; JNKK1; MAP2K4; MAPKK4; MEK4; MKK4; PRKMK4; SAPKK-1; SAPKK1; SEK1; SERK1; SKK1
UniProt ID: (Human) P45985
Entrez Gene ID: (Human) 6416
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.