OriGene
Manufacturer Code:CF505716
Catalog # 50-166-3863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G8 |
Immunogen | Full length human recombit protein of human MAP2K2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CFC4 MAPKK2 MEK2 MKK2 PRKMK2; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; MAP kinase kinase 2; MAP kinase/Erk kinase; MAPK/ERK kinase 2; MAPKK 2; MEK 2; MEK2; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; protein kinase, mitogen activated, kinase 2, p45
Gene Aliases: AA589381; CFC4; MAP2K2; MAPKK2; MEK2; MK2; MKK2; PRKMK2
UniProt ID: (Dog) Q1HG70, (Human) P36507, (Rat) P36506, (Mouse) Q63932
Entrez Gene ID: (Dog) 611939, (Human) 5605, (Rat) 58960, (Mouse) 26396
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.