OriGene
Manufacturer Code:CF500466
Catalog # 50-165-9536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F6 |
Immunogen | Full-length protein expressed in 293T cell transfected with human MAP2K2 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CFC4 MAPKK2 MEK2 MKK2 PRKMK2; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; MAP kinase kinase 2; MAPK/ERK kinase 2; MEK 2; mitogen-activated protein kinase kinase 2, p45
Gene Aliases: CFC4; MAP2K2; MAPKK2; MEK2; MKK2; PRKMK2
UniProt ID: (Human) P36507
Entrez Gene ID: (Human) 5605
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.