Novus Biologicals
Manufacturer Code:NBP147281
Catalog # NBP147281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 475 and 525 of human MAP1S using the numbering given in entry NP_060644.4 (GeneID 55201). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BPY2 interacting protein 1; BPY2 interacting protein 1 BPY2IP1VCY2IP-1 C19orf5 chromosome 19 open reading frame 5 FLJ10669 MAP8MAP-1S MGC133087 microtubule-associated protein 1S Microtubule-associated protein 8 Variable charge Y chromosome 2-interacting protein 1 VCY2 interacting protein 1 VCY2-interacting protein 1 VCY2IP1BPY2-interacting protein 1; BPY2-interacting protein 1; MAP-1S; MAP1S heavy chain; MAP1S light chain; Microtubule-associated protein 1S; Microtubule-associated protein 8; Variable charge Y chromosome 2-interacting protein 1; VCY2 interacting protein 1; VCY2-interacting protein 1
Gene Aliases: BPY2IP1; C19orf5; MAP1S; MAP8; VCY2IP-1; VCY2IP1
UniProt ID: (Human) Q66K74
Entrez Gene ID: (Human) 55201
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.