Invitrogen
Manufacturer Code:MA512816
Catalog # PIMA512816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G5 |
Immunogen | Purified bovine brain MAPs |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MAP-1B; MAP1 light chain LC1; MAP1B heavy chain; Microtubule-associated protein 1B; Neuraxin; protein phosphatase 1, regulatory subunit 102
Gene Aliases: FUTSCH; MAP1B; MAP5; Mtap1b; PPP1R102
UniProt ID: (Human) P46821, (Rat) P15205
Entrez Gene ID: (Bovine) 514739, (Human) 4131, (Rat) 29456
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.