Invitrogen
Manufacturer Code:PA578052
Catalog # PIPA578052
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human MAP1B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A230055D22 AI843217 LC1 MAP5 Mtap1b Mtap5 microtubule-associated protein 1B; MAP-1B; MAP1 light chain LC1; MAP1(X); MAP1.2; MAP1B heavy chain; Microtubule-associated protein 1B; Microtubule-associated protein 1B (MAP 1B) (MAP1.2) (MAP1(X)); microtubule-associated protein 5; Neuraxin; protein phosphatase 1, regulatory subunit 102
Gene Aliases: A230055D22; AI843217; FUTSCH; LC1; MAP1B; MAP5; Mtap-5; Mtap1b; Mtap5; PPP1R102
UniProt ID: (Human) P46821, (Rat) P15205, (Mouse) P14873
Entrez Gene ID: (Human) 4131, (Rat) 29456, (Mouse) 17755
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.