Invitrogen
Manufacturer Code:MA525860
Catalog # PIMA525860
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D6 |
Immunogen | Human recombit protein fragment corresponding to amino acids 249-484 of MAOA produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Amine oxidase [flavin-containing] A; MAO-A; MAOA Amine oxidase &lsqbflavin-containing] A Monoamine oxidase type A MAO-A; Monoamine oxidase type A
Gene Aliases: MAO-A; MAOA
UniProt ID: (Human) P21397
Entrez Gene ID: (Human) 4128
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.