Invitrogen
Manufacturer Code:MA535443
Catalog # PIMA535443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0900 |
Immunogen | A synthesized peptide derived from human Monoamine Oxidase A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110061B18Rik AA407771 Amine oxidase [flavin-containing] A HGNC:6833 Mao Maoa MAO-A monoamine oxidase A Monoamine oxidase type A RP1-201D17__B.2; Amine oxidase [flavin-containing] A; MAO-A; Monoamine oxidase type A
Gene Aliases: 1110061B18Rik; AA407771; Mao; MAO-A; MAOA
UniProt ID: (Human) P21397, (Mouse) Q64133
Entrez Gene ID: (Human) 4128, (Mouse) 17161, (Rat) 29253
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.