Invitrogen
Manufacturer Code:PA586581
Catalog # PIPA586581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 330-380 of Human MAOA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amine oxidase &lsqbflavin-containing] A Monoamine oxidase type A MAO-A MAOA; Amine oxidase [flavin-containing] A; MAO-A; Monoamine oxidase type A
Gene Aliases: 1110061B18Rik; AA407771; Mao; MAO-A; MAOA
UniProt ID: (Human) P21397, (Mouse) Q64133
Entrez Gene ID: (Human) 4128, (Mouse) 17161, (Rat) 29253
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.