Invitrogen
Manufacturer Code:PA569639
Catalog # PIPA569639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human MANEAL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein endo-alpha-1,2-mannosidase-like protein; glycoprotein endo-alpha-12-mannosidase-like protein; mannosidase, endo-alpha-like
Gene Aliases: Gm50; MANEAL; RGD1563973; si:ch211-30b16.2
UniProt ID: (Human) Q5VSG8, (Mouse) Q6P1J0, (Zebrafish) Q1L8D2
Entrez Gene ID: (Bovine) 526978, (Dog) 100855849, (Human) 149175, (Horse) 100069092, (Pig) 110261152, (Rabbit) 100353568, (Guinea pig) 100721941, (Mouse) 215090, (Rat) 366466, (Zebrafish) 566679
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.