Proteintech
Manufacturer Code:212961AP150UL
Catalog # 50-173-3355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MANEA Fusion Protein Ag15847 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 1,2-endomannosidase; ENDO Endo alpha mannosidase Endomannosidase hEndo Mandaselin MANEA mannosidase endo alpha; Endo-alpha mannosidase; endomannosidase; Glycoprotein endo-alpha-1,2-mannosidase; Mandaselin; mannosidase, endo-alpha; mEndo
Gene Aliases: 4932703L02Rik; ENDO; hEndo; MANEA
UniProt ID: (Human) Q5SRI9, (Mouse) Q6NXH2
Entrez Gene ID: (Human) 79694, (Mouse) 242362
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.