Invitrogen
Manufacturer Code:PA531780
Catalog # PIPA531780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 245 of Human MAN2B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,6-mannosidase; core-specific lysosomal alpha-1,6-Mannosidase; core-specific lysosomal alpha-16-Mannosidase epididymis-specific alpha-mannosidase; Epididymis-specific alpha-mannosidase; Mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2
Gene Aliases: KIAA0935; MAN2B2
UniProt ID: (Human) Q9Y2E5
Entrez Gene ID: (Human) 23324
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.