Proteintech
Manufacturer Code:176971AP150UL
Catalog # 50-173-1727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MAN2B2 Fusion Protein Ag11952 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 135 kD alpha-D-mannosidase; 135 kDa alpha-D-mannosidase; Alpha-1,6-mannosidase; core-specific lysosomal alpha-1,6-Mannosidase; Epididymis-specific alpha-mannosidase; KIAA0935 MAN2B2; mannosidase 2, alpha B2; Mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2
Gene Aliases: Ab2-450; KIAA0935; MAN2B2; mKIAA0935
UniProt ID: (Human) Q9Y2E5, (Mouse) O54782
Entrez Gene ID: (Human) 23324, (Mouse) 17160, (Rat) 360955
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.