Abnova
Manufacturer Code:H00023324K
Catalog # 89-209-624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MAN2B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MAN2B2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Alpha-1,6-mannosidase; core-specific lysosomal alpha-1,6-Mannosidase; Epididymis-specific alpha-mannosidase; KIAA0935; Mannosidase alpha class 2B member 2; mannosidase, alpha, class 2B, member 2
Gene Aliases: KIAA0935; MAN2B2
UniProt ID: (Human) Q9Y2E5
Entrez Gene ID: (Human) 23324
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.