Novus Biologicals
Manufacturer Code:NBP183159
Catalog # NBP183159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HHDAITGTAKEAVVVDYGVRLLRSLVNLKQVIIHAAHYLVLGDKETYHFDPEAPFLQVDDTRLSHDALPERTVIQLDSSPRFVVLFNPLEQERFSMVSLLVNSPRVRVLSEEGQPLAVQISAHWSSATEAVPDVYQVSVP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-mannosidase 2x; alpha-mannosidase 2x alpha-mannosidase IIx MAN IIx MANA2X mannosidase alpha type II-X mannosidase alpha class 2A member 2 mannosyl-oligosaccharide 13-16-alpha-mannosidase manosidase alpha- type II isozyme X; Alpha-mannosidase IIx; MAN IIx; Mannosidase alpha class 2A member 2; mannosidase, alpha type II-X; mannosidase, alpha, class 2A, member 2; Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase; manosidase, alpha-, type II, isozyme X
Gene Aliases: MAN2A2; MANA2X
UniProt ID: (Human) P49641
Entrez Gene ID: (Human) 4122
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.