Invitrogen
Manufacturer Code:PA518851
Catalog # PIPA518851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KISSDIKSQNR) corresponding to the internal amino acids of MAN2A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-mannosidase 2; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; MAN2A1 mannosidase alpha class 2A member 1 GOLIM7 MANA2 MANII Golgi alpha-mannosidase II golgi integral membrane protein 7 mannosidase alpha type II mannosyl-oligosaccharide 13-16-alpha-mannosidase; Mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
Gene Aliases: AMan II; GOLIM7; MAN2A1; MANA2; MANII
UniProt ID: (Human) Q16706
Entrez Gene ID: (Human) 4124
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.