Invitrogen
Manufacturer Code:PA5115539
Catalog # PIPA5115539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Mannosidase II corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-mannosidase 2; alpha-mannosidase 2 alpha-mannosidase II AMan II Golgi alpha-mannosidase II golgi integral membrane protein 7 GOLIM7 MAN II MAN2A1 Mana2 Mana-2 MANII Manno mannosidase 2 alpha 1 mannosidase alpha 2 mannosidase alpha class 2A member 1 mannosidase alpha type II mannosidase alpha class 2A member 1 mannosyl-oligosaccharide 13-16-alpha-mannosidase Map-2; alpha-mannosidase II; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; MAN II; mannosidase 2, alpha 1; mannosidase alpha 2; Mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
Gene Aliases: AMan II; GOLIM7; MAN2A1; Mana-2; MANA2; MANII; Manno; Map-2
UniProt ID: (Human) Q16706, (Mouse) P27046, (Rat) P28494
Entrez Gene ID: (Human) 4124, (Mouse) 17158, (Rat) 25478
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.