Invitrogen
Manufacturer Code:PA5112894
Catalog # PIPA5112894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Alpha-mannosidase 2 protein (31-181AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-mannosidase 2; Alpha-mannosidase 2 alpha-mannosidase II AMan II Golgi alpha-mannosidase II golgi integral membrane protein 7 GOLIM7 MAN II Man2a1 MANA2 Mana-2 MANII Manno mannosidase 2 alpha 1 mannosidase alpha 2 Mannosidase alpha class 2A member 1 mannosidase alpha type II mannosidase alpha class 2A member 1 Mannosyl-oligosaccharide 13-16-alpha-mannosidase Map-2; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; Mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
Gene Aliases: AMan II; GOLIM7; MAN2A1; MANA2; MANII
UniProt ID: (Human) Q16706
Entrez Gene ID: (Human) 4124
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.