Novus Biologicals
Manufacturer Code:NBP292006100UL
Catalog # NB025905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 865-1144 of human MAN2A1 (NP_002363.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-mannosidase 2; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; Mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; Mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase
Gene Aliases: AMan II; GOLIM7; MAN2A1; MANA2; MANII
UniProt ID: (Human) Q16706
Entrez Gene ID: (Human) 4124
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.