Invitrogen
Manufacturer Code:PA5104494
Catalog # PIPA5104494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAN1B1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3.2.1.113 E430019H13Rik endoplasmic Reticulum Class I alpha-mannosidase endoplasmic reticulum mannosyl-oligosaccharide 12-alpha-mannosidase endoplasmic reticulum mannosyl-oligosaccharide 12-alpha-mannosidase 1 ER alpha 12-mannosidase ER alpha-12-mannosidase ER mannosidase 1 ERMan1 ERManI Gm108 MAN1B1 Man9GlcNAc2-specific processing alpha-mannosidase Man9GlcNAc2-specific-processing alpha-mannosidase MANA-ER Mannosidase alpha class 1B member 1 mannosidase alpha class 1B member 1 MRT15 RGD1563595 RP11-350O14.2 UNQ747/PRO1477; endoplasmic Reticulum Class I alpha-mannosidase; Endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1; ER alpha 1,2-mannosidase; ER alpha-1,2-mannosidase; ER mannosidase 1; ERMan1; Man9GlcNAc2-specific processing alpha-mannosidase; Man9GlcNAc2-specific-processing alpha-mannosidase; Mannosidase alpha class 1B member 1; mannosidase, alpha, class 1B, member 1
Gene Aliases: E430019H13Rik; ERMAN1; ERManI; Gm108; MAN1B1; MANA-ER; MRT15; RGD1563595; UNQ747/PRO1477
UniProt ID: (Human) Q9UKM7, (Rat) B2GUY0, (Mouse) A2AJ15
Entrez Gene ID: (Human) 11253, (Rat) 499751, (Mouse) 227619
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.