Invitrogen
Manufacturer Code:PA589717
Catalog # PIPA589717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 60-180 of human MAN1A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,2-mannosidase IB; alpha1,2-mannosidase; mannosidase 1, beta; Mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; Processing alpha-1,2-mannosidase IB
Gene Aliases: AI428775; AI528764; AI854422; MAN1A2; MAN1B; PCR2
UniProt ID: (Human) O60476, (Mouse) P39098
Entrez Gene ID: (Human) 10905, (Mouse) 17156
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.