Abnova
Manufacturer Code:H00010905K
Catalog # 89-210-764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MAN1A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MAN1A2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Alpha-1,2-mannosidase IB; alpha1,2-mannosidase; MAN1B; Mannosidase alpha class 1A member 2; mannosidase, alpha, class 1A, member 2; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IB; Processing alpha-1,2-mannosidase IB
Gene Aliases: MAN1A2; MAN1B
UniProt ID: (Human) O60476
Entrez Gene ID: (Human) 10905
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.