Invitrogen
Manufacturer Code:PA569421
Catalog # PIPA569421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human MAN1A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,2-mannosidase IA; Man(9)-alpha-mannosidase; Man9-mannosidase; mannosidase 1, alpha; Mannosidase alpha class 1A member 1; mannosidase, alpha, class 1A, member 1; mannosyl-oligosaccharide 1,2-alpha-mannosidase 1A; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; mannosyl-oligosaccharide 1,2-alpha-mannosidase IA-like; mannosyl-oligosaccharide alpha-1,2-mannosidase; Processing alpha-1,2-mannosidase IA
Gene Aliases: Aa2-166; HUMM3; HUMM9; MAN1A; MAN1A1; MAN9; PCR1; si:dkey-164k19.1; si:dkeyp-6c7.1
UniProt ID: (Human) P33908, (Pig) O02773, (Mouse) P45700
Entrez Gene ID: (Human) 4121, (Pig) 396919, (Rabbit) 100009182, (Dog) 476275, (Bovine) 530027, (Horse) 100072869, (Mouse) 17155, (Rat) 294410, (Zebrafish) 566078
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.