Novus Biologicals
Manufacturer Code:NBP222522
Catalog # NBP222522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to residues C(114)F D G F T Y (R/K) H Y H E N I(126) of human MAL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mal T-cell differentiation protein; mal T-cell differentiation protein myelin and lymphocyte protein T-cell differentiation protein MAL T-lymphocyte maturation-associated protein; MyD88-adapter-like; Myelin and lymphocyte protein; T-lymphocyte maturation-associated protein
Gene Aliases: MAL
UniProt ID: (Human) P21145
Entrez Gene ID: (Human) 4118
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.