Invitrogen
Manufacturer Code:PA561584
Catalog # PIPA561584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MAGOH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mago-nashi homolog, proliferation-associated; MAGOH mago-nashi homolog proliferation-associated (Drosophila) RP5-1024G6.4 MAGOHA; Protein mago nashi homolog
Gene Aliases: MAGOH; MAGOH1; MAGOHA
UniProt ID: (Human) P61326
Entrez Gene ID: (Human) 4116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.