Novus Biologicals
Manufacturer Code:NBP297072R
Catalog # NB041872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant MAGOH (NP_002361.1 Met1-Ile146). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mago-nashi homolog, proliferation-associated; MAGOHAMAGOH1 mago-nashi (Drosophila) homolog proliferation-associated mago-nashi homolog proliferation-associated (Drosophila) protein mago nashi homolog; Protein mago nashi homolog
Gene Aliases: MAGOH; MAGOH1; MAGOHA
UniProt ID: (Human) P61326
Entrez Gene ID: (Human) 4116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.