Abnova
Manufacturer Code:PAB10131
Catalog # 89-114-652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to the guanylate kinase-WW region of MAGI3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Sheep IgG polyclonal antibody raised against partial recombinant MAGI3.
For Research Use Only
Protein Aliases: dJ730K3.2; MAGI-3; Membrane-associated guanylate kinase inverted 3; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 3; membrane-associated guanylate kinase-related 3; MGC163281; RP4-730K3.1
Gene Aliases: dJ730K3.2; KIAA1634; MAGI-3; MAGI3
UniProt ID: (Human) Q5TCQ9
Entrez Gene ID: (Human) 260425
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.