Invitrogen
Manufacturer Code:PA547901
Catalog # PIPA547901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human MAGI2 Ser2-Arg130 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: activin receptor interacting protein 1; Activin receptor-interacting protein 1; Acvrip1; ACVRIP1 AIP-1 AIP1 ARIP1 MAGI-2 SSCAM activin receptor interacting protein 1 atrophin-1-interacting protein 1 atrophin-1-interacting protein A membrane-associated guanylate kinase inverted 2 membrane-associated guanylate kinase WW and PDZ domain-containing protein 2; AIP-1; Atrophin-1-interacting protein 1; Atrophin-1-interacting protein A; MAGI-2; Membrane-associated guanylate kinase inverted 2; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2
Gene Aliases: Acvri1; ACVRINP1; ACVRIP1; AIP-1; AIP1; ARIP1; KIAA0705; MAGI-2; MAGI2; mKIAA0705; S-SCAM; SSCAM
UniProt ID: (Human) Q86UL8, (Mouse) Q9WVQ1
Entrez Gene ID: (Human) 9863, (Mouse) 50791
Molecular Function:
cell junction protein
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.