Invitrogen
Manufacturer Code:PA5101196
Catalog # PIPA5101196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAGI2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin receptor interacting protein 1; activin receptor interacting protein 1 activin receptor-interacting protein 1 Acvri1 Acvrinp1 Acvrip1 AIP1 AIP-1 ARIP1 Atrophin-1-interacting protein 1 atrophin-1-interacting protein A KIAA0705 MAGI2 Magi-2 membrane associated guanylate kinase WW and PDZ domain containing 2 Membrane-associated guanylate kinase inverted 2 membrane-associated guanylate kinase WW and PDZ domain-containing protein 2 mKIAA0705 Sscam S-SCAM Synaptic-scaffolding molecule; Activin receptor-interacting protein 1; Acvrip1; AIP-1; Atrophin-1-interacting protein 1; Atrophin-1-interacting protein A; MAGI-2; Membrane-associated guanylate kinase inverted 2; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2; S-SCAM; Synaptic-scaffolding molecule
Gene Aliases: Acvri1; ACVRINP1; ACVRIP1; AIP-1; AIP1; ARIP1; KIAA0705; MAGI-2; MAGI2; mKIAA0705; S-SCAM; SSCAM
UniProt ID: (Human) Q86UL8, (Rat) O88382, (Mouse) Q9WVQ1
Entrez Gene ID: (Human) 9863, (Rat) 113970, (Mouse) 50791
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.