R&D Systems
Manufacturer Code:AF7117SP
Catalog # AF7117SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human MAGI2 Ser2-Arg130 Accession # Q86UL8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: activin receptor interacting protein 1; ACVRINP1 ACVRIP1 AIP1 AIP1AIP-1 ARIP1 atrophin 1 interacting protein 1 atrophin-1 interacting protein A Atrophin-1-interacting protein 1 Atrophin-1-interacting protein A KIAA0705activin receptor interacting protein 1 MAGI-2SSCAM membrane associated guanylate kinase WW and PDZ domain containing 2 Membrane-associated guanylate kinase inverted 2 membrane-associated guanylate kinase WW and PDZ domain-containing protein 2 SSCAM; AIP-1; Atrophin-1-interacting protein 1; Atrophin-1-interacting protein A; MAGI-2; Membrane-associated guanylate kinase inverted 2; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2
Gene Aliases: ACVRINP1; ACVRIP1; AIP-1; AIP1; ARIP1; KIAA0705; MAGI-2; MAGI2; SSCAM
UniProt ID: (Human) Q86UL8
Entrez Gene ID: (Human) 9863
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.