Abnova
Manufacturer Code:PAB4760
Catalog # 89-131-560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human MAGEH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MAGEH1.
For Research Use Only
Protein Aliases: Apoptosis-related protein 1; APR-1; APR1; MAGE-H1 antigen; MAGEH; melanoma antigen family H, 1; melanoma antigen family H1; Melanoma-associated antigen H1; Restin
Gene Aliases: APR-1; APR1; MAGEH; MAGEH1
UniProt ID: (Human) Q9H213
Entrez Gene ID: (Human) 28986
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.